The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012241.1 | NP_001012241.1 | 5 - 6 | 978 | 109 |
XM_005257298.4 | XP_005257355.1 | 4 - 5 | 2630 | 109 | |
GenBank mRNA | AK125654.1 | - | 1 - 2 | 588 | 109 |
AK297619.1 | - | 2 - 3 | 299 | 109 | |
BC118997.1 | - | 5 - 6 | 901 | 109 | |
BC122543.1 | - | 5 - 6 | 901 | 109 | |
CR749360.1 | - | 5 - 6 | 929 | 109 |