The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012241.1 | NP_001012241.1 | 9 - 10 | 1310 | 73 |
XM_005257298.4 | XP_005257355.1 | 8 - 9 | 2962 | 73 | |
XM_005257299.4 | XP_005257356.3 | 7 - 8 | 2914 | 73 | |
GenBank mRNA | AK124185.1 | - | 3 - 4 | 802 | 73 |
AK125654.1 | - | 5 - 6 | 920 | 73 | |
AK297619.1 | - | 6 - 7 | 631 | 73 | |
BC118997.1 | - | 9 - 10 | 1233 | 73 | |
BC122543.1 | - | 9 - 10 | 1233 | 73 | |
BX647188.1 | - | 3 - 4 | 221 | 73 | |
CR749360.1 | - | 9 - 10 | 1261 | 73 |