The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012353.2 | NP_036485.2 | 1 - 1 | 889 | 115 |
GenBank mRNA | BC140914.1 | - | 1 - 1 | 1161 | 115 |