Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMIM7
Entrez Gene ID
79086
Gene Name
small integral membrane protein 7
Gene Aliases
C19orf42
Chromosome Location
Chr.19: 16630741 - 16660177 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC069279.1 - 5 - 6 588 134

View Full Product Details