Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMYD3
Entrez Gene ID
64754
Gene Name
SET and MYND domain containing 3
Gene Aliases
KMT3E, ZMYND1, ZNFN3A1, bA74P14.1
Chromosome Location
Chr.1: 245749340 - 246507342 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167740.1 NP_001161212.1 6 - 7 716 120
NM_022743.2 NP_073580.1 6 - 7 544 120
XM_011544253.1 XP_011542555.1 6 - 7 692 120
XM_011544254.2 XP_011542556.1 2 - 3 688 120
XM_011544257.1 XP_011542559.1 2 - 3 180 120
XM_011544258.2 XP_011542560.1 3 - 4 537 120
XM_011544259.2 XP_011542561.1 2 - 3 403 120
XM_011544260.1 XP_011542562.1 2 - 3 220 120
XM_011544261.2 XP_011542563.1 2 - 3 145 120
XM_017002094.1 XP_016857583.1 2 - 3 4360 120
GenBank mRNA AB057595.1 - 6 - 7 686 120
AK023594.1 - 1 - 2 946 120
AK024733.1 - 6 - 7 541 120
AK289605.1 - 6 - 7 692 120
AY186742.1 - 3 - 4 455 120
BC017079.1 - 2 - 3 168 120
BC031010.1 - 6 - 7 631 120

View Full Product Details