The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199692.1 | NP_001186621.1 | 8 - 9 | 2183 | 137 |
NM_001199693.1 | NP_001186622.1 | 7 - 8 | 1209 | 137 | |
NM_001199694.1 | NP_001186623.1 | 7 - 8 | 1630 | 137 | |
NM_003040.3 | NP_003031.3 | 8 - 9 | 1399 | 137 | |
XM_006716094.3 | XP_006716157.1 | 8 - 9 | 1399 | 137 | |
GenBank mRNA | AB209158.1 | - | 8 - 9 | 2183 | 137 |
AK295767.1 | - | 7 - 8 | 1209 | 137 | |
AK312699.1 | - | 8 - 9 | 1378 | 137 | |
BC004893.2 | - | 8 - 9 | 1036 | 137 | |
BC009386.2 | - | 7 - 8 | 1630 | 137 | |
BC009434.1 | - | 7 - 8 | 1630 | 137 | |
BC010069.2 | - | 8 - 9 | 1036 | 137 | |
BC028601.2 | - | 7 - 8 | 1231 | 137 | |
DA699325.1 | - | 1 - 2 | 158 | 137 | |
U62531.1 | - | 8 - 9 | 1317 | 137 | |
X62137.1 | - | 8 - 9 | 1222 | 137 |