The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003816.2 | NP_003807.1 | 18 - 19 | 2145 | 101 |
XM_011544682.2 | XP_011542984.1 | 18 - 19 | 2395 | 101 | |
XM_017013942.1 | XP_016869431.1 | 18 - 19 | 2088 | 101 | |
GenBank mRNA | AK293257.1 | - | 5 - 6 | 511 | 101 |
D14665.2 | - | 14 - 15 | 1720 | 101 | |
U41766.1 | - | 18 - 19 | 2145 | 101 |