Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VEZT
Entrez Gene ID
55591
Gene Name
vezatin, adherens junctions transmembrane protein
Gene Aliases
VEZATIN
Chromosome Location
Chr.12: 95217746 - 95302799 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017599.3 NP_060069.3 6 - 7 958 130
NR_038241.1 7 - 8 1031 130
NR_038242.1 6 - 7 966 130
XM_006719477.3 XP_006719540.1 6 - 7 970 130
XM_006719478.2 XP_006719541.1 5 - 6 864 130
XM_006719479.2 XP_006719542.1 7 - 8 1041 130
XM_006719480.3 XP_006719543.1 7 - 8 1018 130
XM_006719481.2 XP_006719544.1 6 - 7 888 130
XM_006719482.3 XP_006719545.1 6 - 7 1269 130
XM_006719483.3 XP_006719546.1 6 - 7 1286 130
XM_006719484.3 XP_006719547.1 5 - 6 1196 130
XM_006719485.2 XP_006719548.1 4 - 5 638 130
XM_006719486.3 XP_006719549.1 4 - 5 626 130
XM_006719490.3 XP_006719553.1 7 - 8 1029 130
XM_011538543.1 XP_011536845.1 7 - 8 963 130
XM_011538544.1 XP_011536846.1 7 - 8 951 130
XM_011538545.2 XP_011536847.1 6 - 7 931 130
XM_011538546.1 XP_011536848.1 8 - 9 1034 130
XM_011538547.1 XP_011536849.1 8 - 9 1078 130
XM_011538548.2 XP_011536850.1 5 - 6 724 130
XM_011538549.1 XP_011536851.1 7 - 8 963 130
XM_011538550.2 XP_011536852.1 7 - 8 963 130
XM_011538551.2 XP_011536853.1 7 - 8 951 130
XM_011538552.1 XP_011536854.1 7 - 8 1037 130
XM_011538553.2 XP_011536855.1 7 - 8 967 130
XM_017019586.1 XP_016875075.1 6 - 7 919 130
XM_017019587.1 XP_016875076.1 5 - 6 852 130
XM_017019588.1 XP_016875077.1 8 - 9 1022 130
XM_017019589.1 XP_016875078.1 7 - 8 1006 130
XM_017019590.1 XP_016875079.1 8 - 9 1066 130
XM_017019591.1 XP_016875080.1 6 - 7 1257 130
XM_017019592.1 XP_016875081.1 5 - 6 711 130
XM_017019593.1 XP_016875082.1 8 - 9 1086 130
XM_017019594.1 XP_016875083.1 7 - 8 948 130
XM_017019595.1 XP_016875084.1 6 - 7 970 130
XM_017019596.1 XP_016875085.1 6 - 7 970 130
XM_017019597.1 XP_016875086.1 6 - 7 958 130
XM_017019598.1 XP_016875087.1 6 - 7 958 130
XM_017019599.1 XP_016875088.1 6 - 7 919 130
XM_017019600.1 XP_016875089.1 6 - 7 1274 130
XM_017019601.1 XP_016875090.1 7 - 8 1025 130
XM_017019602.1 XP_016875091.1 8 - 9 1022 130
XM_017019603.1 XP_016875092.1 6 - 7 1286 130
XM_017019604.1 XP_016875093.1 4 - 5 638 130
XM_017019605.1 XP_016875094.1 4 - 5 626 130
GenBank mRNA AF216644.1 - 6 - 7 885 130
AF225417.1 - 6 - 7 929 130
AF277625.1 - 2 - 3 397 130
AK001338.1 - 4 - 5 639 130
AK225303.1 - 7 - 8 971 130
AK225356.1 - 7 - 8 980 130
AK307649.1 - 7 - 8 1037 130
AK309943.1 - 6 - 7 905 130
AL137497.1 - 7 - 8 953 130
BC064939.1 - 7 - 8 948 130

View Full Product Details