Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VEZT
Entrez Gene ID
55591
Gene Name
vezatin, adherens junctions transmembrane protein
Gene Aliases
VEZATIN
Chromosome Location
Chr.12: 95217746 - 95302799 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017599.3 NP_060069.3 8 - 9 1431 151
NR_038241.1 9 - 10 1504 151
NR_038242.1 8 - 9 1439 151
XM_006719477.3 XP_006719540.1 8 - 9 1443 151
XM_006719478.2 XP_006719541.1 7 - 8 1337 151
XM_006719479.2 XP_006719542.1 9 - 10 1514 151
XM_006719480.3 XP_006719543.1 9 - 10 1491 151
XM_006719481.2 XP_006719544.1 8 - 9 1361 151
XM_006719482.3 XP_006719545.1 8 - 9 1742 151
XM_006719483.3 XP_006719546.1 8 - 9 1759 151
XM_006719484.3 XP_006719547.1 7 - 8 1669 151
XM_006719485.2 XP_006719548.1 6 - 7 1111 151
XM_006719486.3 XP_006719549.1 6 - 7 1099 151
XM_006719490.3 XP_006719553.1 9 - 10 1502 151
XM_011538543.1 XP_011536845.1 9 - 10 1436 151
XM_011538544.1 XP_011536846.1 9 - 10 1424 151
XM_011538545.2 XP_011536847.1 8 - 9 1404 151
XM_011538546.1 XP_011536848.1 10 - 11 1507 151
XM_011538547.1 XP_011536849.1 10 - 11 1551 151
XM_011538548.2 XP_011536850.1 7 - 8 1197 151
XM_011538549.1 XP_011536851.1 9 - 10 1436 151
XM_011538550.2 XP_011536852.1 9 - 10 1436 151
XM_011538551.2 XP_011536853.1 9 - 10 1424 151
XM_011538552.1 XP_011536854.1 9 - 10 1510 151
XM_011538553.2 XP_011536855.1 9 - 10 1440 151
XM_017019586.1 XP_016875075.1 8 - 9 1392 151
XM_017019587.1 XP_016875076.1 7 - 8 1325 151
XM_017019588.1 XP_016875077.1 10 - 11 1495 151
XM_017019589.1 XP_016875078.1 9 - 10 1479 151
XM_017019590.1 XP_016875079.1 10 - 11 1539 151
XM_017019591.1 XP_016875080.1 8 - 9 1730 151
XM_017019592.1 XP_016875081.1 7 - 8 1184 151
XM_017019593.1 XP_016875082.1 10 - 11 1559 151
XM_017019594.1 XP_016875083.1 9 - 10 1421 151
XM_017019595.1 XP_016875084.1 8 - 9 1443 151
XM_017019596.1 XP_016875085.1 8 - 9 1443 151
XM_017019597.1 XP_016875086.1 8 - 9 1431 151
XM_017019598.1 XP_016875087.1 8 - 9 1431 151
XM_017019599.1 XP_016875088.1 8 - 9 1392 151
XM_017019600.1 XP_016875089.1 8 - 9 1747 151
XM_017019601.1 XP_016875090.1 9 - 10 1498 151
XM_017019602.1 XP_016875091.1 10 - 11 1495 151
XM_017019603.1 XP_016875092.1 8 - 9 1759 151
XM_017019604.1 XP_016875093.1 6 - 7 1111 151
XM_017019605.1 XP_016875094.1 6 - 7 1099 151
GenBank mRNA AF216644.1 - 8 - 9 1358 151
AF225417.1 - 8 - 9 1402 151
AF277625.1 - 4 - 5 870 151
AK225303.1 - 9 - 10 1444 151
AK225356.1 - 9 - 10 1453 151
AK307649.1 - 9 - 10 1510 151
AK309943.1 - 8 - 9 1378 151
AL133113.1 - 1 - 2 157 151
AL137497.1 - 9 - 10 1426 151
BC028597.1 - 11 - 12 1760 151
BC064939.1 - 9 - 10 1421 151

View Full Product Details