The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166412.1 | NP_001159884.1 | 10 - 11 | 1228 | 126 |
NM_022138.2 | NP_071421.1 | 10 - 11 | 1261 | 126 | |
XM_011536065.1 | XP_011534367.1 | 10 - 11 | 1259 | 126 | |
XM_011536066.1 | XP_011534368.1 | 10 - 11 | 1226 | 126 | |
GenBank mRNA | AB014730.1 | - | 10 - 11 | 1028 | 126 |
AB014737.1 | - | 10 - 11 | 1061 | 126 | |
AF173892.1 | - | 9 - 10 | 883 | 126 | |
AJ249902.1 | - | 5 - 6 | 507 | 126 | |
AJ420521.1 | - | 10 - 11 | 1208 | 126 | |
AK056700.1 | - | 10 - 11 | 1155 | 126 | |
AK297839.1 | - | 2 - 3 | 153 | 126 | |
AK314613.1 | - | 10 - 11 | 1195 | 126 | |
AK315920.1 | - | 2 - 3 | 517 | 126 | |
AL832303.1 | - | 10 - 11 | 1163 | 126 | |
BC028420.1 | - | 10 - 11 | 1231 | 126 | |
BC047583.1 | - | 10 - 11 | 1200 | 126 |