Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMOC2
Entrez Gene ID
64094
Gene Name
SPARC related modular calcium binding 2
Gene Aliases
DTDP1, MST117, MSTP117, MSTP140, SMAP2, bA270C4A.1, bA37D8.1, dJ421D16.1
Chromosome Location
Chr.6: 168441151 - 168667994 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166412.1 NP_001159884.1 9 - 10 1130 115
NM_022138.2 NP_071421.1 9 - 10 1163 115
XM_011536065.1 XP_011534367.1 9 - 10 1161 115
XM_011536066.1 XP_011534368.1 9 - 10 1128 115
GenBank mRNA AB014730.1 - 9 - 10 930 115
AB014737.1 - 9 - 10 963 115
AF173892.1 - 8 - 9 785 115
AJ249902.1 - 4 - 5 409 115
AJ420521.1 - 9 - 10 1110 115
AK056700.1 - 9 - 10 1057 115
AK314613.1 - 9 - 10 1097 115
AL832303.1 - 9 - 10 1065 115
BC028420.1 - 9 - 10 1133 115
BC047583.1 - 9 - 10 1102 115

View Full Product Details