The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159676.1 | NP_001153148.1 | 10 - 11 | 1656 | 178 |
XM_005248635.4 | XP_005248692.1 | 11 - 12 | 1806 | 178 | |
XM_017010179.1 | XP_016865668.1 | 11 - 12 | 1741 | 178 | |
XM_017010180.1 | XP_016865669.1 | 10 - 11 | 1821 | 178 |