Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNM1P35
Entrez Gene ID
100128285
Gene Name
dynamin 1 pseudogene 35
Gene Aliases
DNM1DN8-2, DNM1DN8.2, DNM1DN8@, FKSG88
Chromosome Location
Chr.15: 75727670 - 75740077 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_024595.3 4 - 5 798 174
GenBank mRNA AK091244.1 - 2 - 3 419 174

View Full Product Details