Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C7orf72
Entrez Gene ID
100130988
Gene Name
chromosome 7 open reading frame 72
Gene Aliases
-
Chromosome Location
Chr.7: 50095861 - 50159256 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161834.2 NP_001155306.2 3 - 4 723 100
XM_011515052.1 XP_011513354.1 3 - 4 948 100
XM_011515053.2 XP_011513355.1 3 - 4 948 100
GenBank mRNA BX092263.1 - 3 - 4 274 100

View Full Product Details