Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASP8AP2
Entrez Gene ID
9994
Gene Name
caspase 8 associated protein 2
Gene Aliases
CED-4, FLASH, RIP25
Chromosome Location
Chr.6: 89829900 - 89874436 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001137667.1 NP_001131139.1 1 - 2 165 174
GenBank mRNA BP363682.1 - 1 - 2 165 174

View Full Product Details