Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRHR1-IT1
Entrez Gene ID
147081
Gene Name
CRHR1 intronic transcript 1
Gene Aliases
C17orf69
Chromosome Location
Chr.17: 45638975 - 45646229 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_026906.2 2 - 3 611 166


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG187386.1 - 2 - 3 213 Chr. 17 - 45639374 - 45646232 [+] on Build GRCh38 166
GenBankmRNA AI299973.1 - 1 - 2 Chr. 17 - 45640389 - 45646227 [-] on Build GRCh38 166
GenBankmRNA BE042948.1 - 1 - 2 Chr. 17 - 45639407 - 45646227 [-] on Build GRCh38 166
GenBankmRNA BF448276.1 - 1 - 2 Chr. 17 - 45639437 - 45646232 [-] on Build GRCh38 166
GenBankmRNA BF223398.1 - 1 - 2 Chr. 17 - 45639394 - 45646226 [-] on Build GRCh38 166


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream MGC57346-CRHR1 readthrough MGC57346-CRHR1 Anti-sense Chr. - 45620329 - 45835828 on Build GRCh38
Downstream corticotropin releasing hormone receptor 1 CRHR1 CRF-R, CRF-R-1, CRF-R1, CRF1, CRFR-1, CRFR1, CRH-R-1, CRH-R1, CRHR, CRHR1L 138.1kb Anti-sense Chr. - 45784280 - 45835828 on Build GRCh38

View Full Product Details