Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRAP
Entrez Gene ID
4892
Gene Name
nebulin related anchoring protein
Gene Aliases
N-RAP
Chromosome Location
Chr.10: 113588824 - 113664070 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261463.1 NP_001248392.1 1 - 2 265 88
NM_001322945.1 NP_001309874.1 1 - 2 265 88
NM_006175.4 NP_006166.3 1 - 2 265 88
NM_198060.3 NP_932326.2 1 - 2 265 88
XM_005269864.2 XP_005269921.1 1 - 2 241 88
XM_005269865.2 XP_005269922.1 1 - 2 241 88
XM_005269867.2 XP_005269924.1 1 - 2 241 88
XM_006717870.2 XP_006717933.1 1 - 2 241 88
XM_011539832.2 XP_011538134.1 1 - 2 241 88
GenBank mRNA AK056969.1 - 1 - 2 242 88
AL831980.2 - 1 - 2 192 88
AL832025.2 - 1 - 2 192 88
AL832330.2 - 1 - 2 192 88
AY081943.2 - 1 - 2 242 88
AY177620.1 - 1 - 2 218 88
AY177621.1 - 1 - 2 218 88
BC005933.1 - 1 - 2 236 88
BC017866.1 - 1 - 2 236 88
BC058825.1 - 1 - 2 241 88
BC070130.1 - 1 - 2 236 88
BC126407.1 - 1 - 2 101 88
BC143751.1 - 1 - 2 101 88
BX640682.2 - 1 - 2 192 88
BX640730.1 - 1 - 2 349 88
BX641052.1 - 1 - 2 229 88
BX647658.1 - 1 - 2 264 88
DA564310.1 - 1 - 2 268 88

View Full Product Details