Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRAP
Entrez Gene ID
4892
Gene Name
nebulin related anchoring protein
Gene Aliases
N-RAP
Chromosome Location
Chr.10: 113588824 - 113664070 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261463.1 NP_001248392.1 34 - 35 4104 106
NM_001322945.1 NP_001309874.1 33 - 34 3996 106
NM_006175.4 NP_006166.3 33 - 34 3999 106
NM_198060.3 NP_932326.2 34 - 35 4104 106
XM_005269864.2 XP_005269921.1 33 - 34 3975 106
XM_005269865.2 XP_005269922.1 33 - 34 3972 106
XM_006717870.2 XP_006717933.1 33 - 34 3975 106
XM_011539832.2 XP_011538134.1 34 - 35 4080 106
GenBank mRNA AL831980.2 - 34 - 35 4031 106
AL832025.2 - 34 - 35 4031 106
AL832330.2 - 34 - 35 4031 106
AL832457.3 - 20 - 21 2457 106
AY081943.2 - 33 - 34 3976 106
AY177620.1 - 34 - 35 4057 106
AY177621.1 - 33 - 34 3952 106
BC126407.1 - 34 - 35 3940 106
BC143751.1 - 34 - 35 3940 106
BX640682.2 - 34 - 35 4031 106
BX640730.1 - 34 - 35 4188 106
BX641052.1 - 34 - 35 4068 106
BX647658.1 - 34 - 35 4105 106

View Full Product Details