The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017825.2 | NP_060295.1 | 3 - 4 | 571 | 164 |
XM_011541636.2 | XP_011539938.1 | 2 - 3 | 441 | 164 | |
GenBank mRNA | AF212236.1 | - | 3 - 4 | 569 | 164 |
AJ427295.1 | - | 3 - 4 | 548 | 164 | |
AK000453.1 | - | 3 - 4 | 538 | 164 | |
AK223037.1 | - | 3 - 4 | 522 | 164 | |
AK295283.1 | - | 4 - 5 | 497 | 164 | |
AK316349.1 | - | 4 - 5 | 484 | 164 | |
BC014169.2 | - | 3 - 4 | 533 | 164 | |
CR457237.1 | - | 3 - 4 | 518 | 164 |