Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PABPC1
- Entrez Gene ID
-
26986
- Gene Name
- poly(A) binding protein cytoplasmic 1
- Gene Aliases
-
PAB1, PABP, PABP1, PABPC2, PABPL1
- Chromosome Location
-
Chr.8: 100702916 - 100722998 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
Y00345.1 |
- |
4 - 5 |
1133 |
140 |
View Full Product Details