The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198560.2 | NP_940962.1 | 2 - 3 | 693 | 70 |
XM_017006359.1 | XP_016861848.1 | 1 - 2 | 1231 | 70 | |
GenBank mRNA | AY278320.1 | - | 2 - 3 | 693 | 70 |
BC113964.1 | - | 2 - 3 | 429 | 70 |