Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRR27
Entrez Gene ID
401137
Gene Name
proline rich 27
Gene Aliases
C4orf40
Chromosome Location
Chr.4: 70154187 - 70166609 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214711.3 NP_999876.2 2 - 3 262 158
GenBank mRNA BX484937.1 - 2 - 3 263 158
BX485867.1 - 2 - 3 259 158
BX640892.1 - 2 - 3 175 158
BX640924.1 - 2 - 3 213 158
BX640997.1 - 2 - 3 249 158
BX647183.1 - 2 - 3 134 158
BX647185.1 - 3 - 4 252 204
BX648724.1 - 2 - 3 211 158
BX648872.1 - 2 - 3 134 158
BX648895.1 - 2 - 3 211 158
BX648896.1 - 3 - 4 146 158

View Full Product Details