The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142418.1 | NP_001135890.1 | 5 - 5 | 1370 | 153 |
NM_001142419.1 | NP_001135891.1 | 4 - 4 | 1303 | 153 | |
NM_001142420.1 | NP_001135892.1 | 3 - 3 | 1150 | 153 | |
NM_001142421.1 | NP_001135893.1 | 4 - 4 | 1252 | 153 | |
NM_001142422.1 | NP_001135894.1 | 5 - 5 | 1352 | 153 | |
NM_001142423.1 | NP_001135895.1 | 4 - 4 | 1270 | 153 | |
NM_001142424.1 | NP_001135896.1 | 5 - 5 | 1319 | 153 | |
NM_001142425.1 | NP_001135897.1 | 4 - 4 | 1290 | 153 | |
NM_001142426.1 | NP_001135898.1 | 3 - 3 | 1168 | 153 | |
NM_001142427.1 | NP_001135899.1 | 4 - 4 | 1272 | 153 | |
NM_001142428.1 | NP_001135900.1 | 5 - 5 | 1301 | 153 | |
NM_001142429.1 | NP_001135901.1 | 5 - 5 | 1339 | 153 | |
NM_001142430.1 | NP_001135902.1 | 4 - 4 | 1217 | 153 | |
NM_001142431.1 | NP_001135903.1 | 4 - 4 | 1199 | 153 | |
NM_001142432.1 | NP_001135904.1 | 5 - 5 | 1321 | 153 | |
NM_012286.2 | NP_036418.1 | 4 - 4 | 1321 | 153 | |
GenBank mRNA | AF100620.1 | - | 4 - 4 | 1171 | 153 |
AF167174.1 | - | 4 - 4 | 1166 | 153 | |
AK056012.1 | - | 4 - 4 | 1158 | 153 | |
AK095298.1 | - | 2 - 2 | 2083 | 153 | |
AK126115.1 | - | 6 - 6 | 1146 | 153 | |
AK126427.1 | - | 4 - 4 | 2098 | 153 | |
AK223504.1 | - | 4 - 4 | 1202 | 153 | |
AK302083.1 | - | 5 - 5 | 1102 | 153 | |
BC056899.1 | - | 5 - 5 | 1156 | 153 | |
BC093013.1 | - | 5 - 5 | 1200 | 153 | |
D14812.1 | - | 4 - 4 | 1171 | 153 |