The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017002849.1 | XP_016858338.1 | 1 - 1 | 3096 | 103 |
XM_017002850.1 | XP_016858339.1 | 1 - 1 | 3095 | 103 | |
XM_017002851.1 | XP_016858340.1 | 1 - 1 | 3095 | 103 | |
XM_017002852.1 | XP_016858341.1 | 1 - 1 | 3094 | 103 |