Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDR1
Entrez Gene ID
1038
Gene Name
cerebellar degeneration related protein 1
Gene Aliases
CDR, CDR34, CDR62A
Chromosome Location
Chr.X: 140783260 - 140784558 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004065.2 NP_004056.2 1 - 1 366 134
GenBank mRNA BC113472.1 - 1 - 1 291 134
BC113474.1 - 1 - 1 291 134
M16965.1 - 1 - 1 234 134

View Full Product Details