The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004457.1 | NP_001004457.1 | 1 - 1 | 868 | 116 |
GenBank mRNA | BC137182.1 | - | 1 - 1 | 926 | 116 |
BC137183.1 | - | 1 - 1 | 926 | 116 |