Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR5H2
Entrez Gene ID
79310
Gene Name
olfactory receptor family 5 subfamily H member 2
Gene Aliases
OR3-10
Chromosome Location
Chr.3: 98282888 - 98283832 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005482.1 NP_001005482.1 1 - 1 813 123

View Full Product Details