The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000693.3 | NP_000684.2 | 13 - 13 | 3140 | 105 |
NM_001293815.1 | NP_001280744.1 | 10 - 10 | 2819 | 105 | |
GenBank mRNA | BC069274.1 | - | 13 - 13 | 2980 | 105 |
BX538027.1 | - | 10 - 10 | 2637 | 105 | |
U07919.1 | - | 13 - 13 | 2976 | 105 |