The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002118.4 | NP_002109.2 | 6 - 6 | 1333 | 124 |
GenBank mRNA | AB209577.1 | - | 2 - 2 | 4505 | 124 |
AY645722.1 | - | 6 - 6 | 1303 | 124 | |
BC027175.1 | - | 6 - 6 | 1214 | 124 | |
CD742571.1 | - | 1 - 1 | 124 | ||
U15085.1 | - | 6 - 6 | 1303 | 124 | |
Z23139.1 | - | 6 - 6 | 1238 | 124 |