The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199279.1 | NP_001186208.1 | 5 - 5 | 2270 | 96 |
NM_004198.3 | NP_004189.1 | 6 - 6 | 2315 | 96 | |
GenBank mRNA | BC014456.1 | - | 6 - 6 | 2005 | 96 |