Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GID4
Entrez Gene ID
79018
Gene Name
GID complex subunit 4 homolog
Gene Aliases
C17orf39, VID24
Chromosome Location
Chr.17: 18039297 - 18068405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024052.4 NP_076957.3 6 - 6 3533 133
XM_005256800.1 XP_005256857.1 5 - 5 3289 133
GenBank mRNA AK124377.1 - 2 - 2 1471 133
AK127580.1 - 6 - 6 3224 133

View Full Product Details