Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BTN2A2
- Entrez Gene ID
-
10385
- Gene Name
- butyrophilin subfamily 2 member A2
- Gene Aliases
-
BT2.2, BTF2, BTN2.2
- Chromosome Location
-
Chr.6: 26382891 - 26394874 on Build GRCh38
View Full Product Details