The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031804.2 | NP_001026974.1 | 1 - 1 | 6193 | 112 |
GenBank mRNA | BC081542.1 | - | 1 - 1 | 5486 | 112 |