Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OTOA
Entrez Gene ID
146183
Gene Name
otoancorin
Gene Aliases
CT108, DFNB22
Chromosome Location
Chr.16: 21663857 - 21771468 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144672.3 NP_653273.3 3 - 4 157 81
XM_011545747.2 XP_011544049.1 4 - 5 536 81
XM_017022950.1 XP_016878439.1 4 - 5 260 81
GenBank mRNA BC129992.1 - 3 - 4 154 81
BC129993.1 - 3 - 4 161 81

View Full Product Details