The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039503.2 | NP_001034592.1 | 9 - 10 | 1422 | 108 |
XM_011545817.1 | XP_011544119.1 | 8 - 9 | 2789 | 108 | |
XM_011545818.2 | XP_011544120.1 | 8 - 9 | 2789 | 108 | |
XM_011545820.1 | XP_011544122.1 | 9 - 10 | 2648 | 108 | |
GenBank mRNA | AJ627035.1 | - | 9 - 10 | 1422 | 108 |