Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCDC1
Entrez Gene ID
341019
Gene Name
doublecortin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.11: 31262624 - 31369810 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181807.3 NP_861523.2 6 - 7 989 95
GenBank mRNA AY247970.1 - 6 - 7 953 95

View Full Product Details