Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXF2
Entrez Gene ID
2295
Gene Name
forkhead box F2
Gene Aliases
FKHL6, FREAC-2, FREAC2
Chromosome Location
Chr.6: 1389834 - 1395597 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001452.1 NP_001443.1 1 - 2 1283 80
GenBank mRNA U13220.1 - 1 - 2 1283 80

View Full Product Details