Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPS29
Entrez Gene ID
6235
Gene Name
ribosomal protein S29
Gene Aliases
DBA13, S29
Chromosome Location
Chr.14: 49576672 - 49586416 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030001.2 NP_001025172.1 2 - 3 229 66
GenBank mRNA AK307833.1 - 2 - 3 190 66
BC015974.1 - 2 - 3 193 66
BF128896.1 - 2 - 3 136 66
BQ433530.1 - 2 - 3 173 66

View Full Product Details