The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076780.1 | NP_001070248.1 | 6 - 7 | 1137 | 88 |
NM_052892.3 | NP_443124.3 | 6 - 7 | 1137 | 88 | |
NR_126532.1 | 6 - 7 | 1137 | 88 | ||
GenBank mRNA | AY164483.1 | - | 6 - 7 | 1137 | 88 |