Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C4orf51
Entrez Gene ID
646603
Gene Name
chromosome 4 open reading frame 51
Gene Aliases
-
Chromosome Location
Chr.4: 145680103 - 145792248 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080531.1 NP_001074000.1 1 - 2 226 77
XM_006714289.3 XP_006714352.1 1 - 2 327 77
XM_011532196.2 XP_011530498.1 1 - 2 327 77
XM_011532197.2 XP_011530499.1 1 - 2 327 77
XM_017008547.1 XP_016864036.1 1 - 2 327 77
XM_017008548.1 XP_016864037.1 1 - 2 327 77
XM_017008549.1 XP_016864038.1 1 - 2 327 77
XM_017008550.1 XP_016864039.1 1 - 2 327 77
XM_017008551.1 XP_016864040.1 1 - 2 327 77
XM_017008552.1 XP_016864041.1 1 - 2 327 77
XM_017008553.1 XP_016864042.1 1 - 2 327 77
XM_017008554.1 XP_016864043.1 1 - 2 327 77
XM_017008555.1 XP_016864044.1 1 - 2 327 77
XM_017008556.1 XP_016864045.1 1 - 2 327 77

View Full Product Details