Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC2A14
Entrez Gene ID
144195
Gene Name
solute carrier family 2 member 14
Gene Aliases
GLUT14, SLC2A3P3
Chromosome Location
Chr.12: 7812512 - 7891196 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286233.1 NP_001273162.1 7 - 8 1061 85
NM_153449.3 NP_703150.1 3 - 4 343 85
GenBank mRNA AF481879.1 - 3 - 4 203 85
AK126026.1 - 7 - 8 843 85
DB069339.1 - 3 - 4 345 85

View Full Product Details