The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098637.1 | NP_001092107.1 | 2 - 3 | 1517 | 86 |
GenBank mRNA | BG260395.1 | - | 1 - 2 | 520 | 86 |