The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099218.2 | NP_001092688.1 | 1 - 2 | 3274 | 136 |
NM_001321233.1 | NP_001308162.1 | 5 - 6 | 3632 | 136 | |
GenBank mRNA | AK310498.1 | - | 6 - 7 | 3879 | 136 |
DQ860102.1 | - | 1 - 2 | 3274 | 136 |