The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102562.1 | NP_001096032.1 | 3 - 4 | 1097 | 101 |
GenBank mRNA | BC150513.1 | - | 1 - 2 | 226 | 101 |
BC150514.1 | - | 1 - 2 | 227 | 101 |