Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
CFHR5 | NM_030787.3 | NP_110414.1 | ||
XM_011510020.2 | XP_011508322.1 | |||
AF295327.1 | AAK15619.1 | |||
AK314026.1 | ||||
BC111773.1 | AAI11774.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2676788 | Chr1:196987254 - 197007832 on Build GRCh38 | Deletion | CFHR5 |
dgv12e55 | Chr1:196762018 - 197042447 on Build GRCh38 | Gain | CFHR5 CFHR3 CFHR4 CFHR2 F13B CFHR1 |
esv2758990 | Chr1:196621707 - 197200642 on Build GRCh38 | Gain+Loss | CFH CFHR5 CFHR3 CFHR4 ASPM CFHR2 F13B CFHR1 ZBTB41 |
nsv528460 | Chr1:196573437 - 197477572 on Build GRCh38 | Gain | CFH CFHR5 CFHR3 CFHR4 KCNT2 CRB1 ASPM CFHR2 F13B CFHR1 MIR4735 ZBTB41 |
dgv72e214 | Chr1:196987015 - 197011691 on Build GRCh38 | Loss | CFHR5 |