Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNORD95
Entrez Gene ID
619570
Gene Name
small nucleolar RNA, C/D box 95
Gene Aliases
U95
Chromosome Location
Chr.5: 181243314 - 181243376 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_002591.1 1 - 1 34 61


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BP312222.1 - 2 280 Chr. 5 - 181243042 - 181243908 [-] on Build GRCh38 61
GenBankmRNA DA524279.1 - 2 276 Chr. 5 - 181243055 - 181243904 [-] on Build GRCh38 61
GenBankmRNA BP214324.1 - 2 238 Chr. 5 - 181241491 - 181243866 [-] on Build GRCh38 61
GenBankmRNA BG820132.1 - 1 56 Chr. 5 - 181243021 - 181243398 [-] on Build GRCh38 61
GenBankmRNA AV735346.1 - 1 81 Chr. 5 - 181242952 - 181243381 [-] on Build GRCh38 61
GenBankmRNA DA510742.1 - 1 520 Chr. 5 - 181243309 - 181243861 [-] on Build GRCh38 61
GenBankmRNA DA077159.1 - 2 276 Chr. 5 - 181242250 - 181243904 [-] on Build GRCh38 61
GenBankmRNA DA273363.1 - 1 431 Chr. 5 - 181243222 - 181243773 [-] on Build GRCh38 61
GenBankmRNA DW420712.1 - 1 52 Chr. 5 - 181243095 - 181243379 [-] on Build GRCh38 61
GenBankmRNA DA376440.1 - 1 398 Chr. 5 - 181243173 - 181243740 [-] on Build GRCh38 61
GenBankmRNA DA074472.1 - 2 280 Chr. 5 - 181243057 - 181243908 [-] on Build GRCh38 61
GenBankmRNA DW453506.1 - 52 Chr. - - [-] on Build 61
GenBankmRNA BG829304.1 - 2 267 Chr. 5 - 181239539 - 181243894 [-] on Build GRCh38 61
GenBankmRNA DW456821.1 - 52 Chr. - - [-] on Build 61
GenBankmRNA DB023326.1 - 2 276 Chr. 5 - 181242215 - 181243904 [-] on Build GRCh38 61
GenBankmRNA DA091684.1 - 2 278 Chr. 5 - 181243058 - 181243906 [-] on Build GRCh38 61


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream small nucleolar RNA, C/D box 96A SNORD96A U96A -1.4kb Sense Chr. - 181241818 - 181241889 on Build GRCh38
Downstream uncharacterized LOC101928649 CTC-338M12.4 3.2kb Anti-sense Chr. - 181246541 - 181257586 on Build GRCh38

View Full Product Details