The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134941.2 | NP_001128413.1 | 2 - 3 | 842 | 50 |
NM_001302821.1 | NP_001289750.1 | 3 - 4 | 593 | 50 | |
NM_001302822.1 | NP_001289751.1 | 2 - 3 | 381 | 50 | |
NM_001302823.1 | NP_001289752.1 | 2 - 3 | 381 | 50 | |
NM_001302824.1 | NP_001289753.1 | 2 - 3 | 303 | 50 | |
NM_016362.4 | NP_057446.1 | 2 - 3 | 842 | 50 | |
XM_017006612.1 | XP_016862101.1 | 1 - 2 | 1377 | 50 | |
XM_017006613.1 | XP_016862102.1 | 1 - 2 | 1386 | 50 | |
GenBank mRNA | AY359053.1 | - | 1 - 2 | 146 | 50 |
EF549567.1 | - | 1 - 2 | 146 | 50 | |
EF549569.1 | - | 1 - 2 | 558 | 50 | |
EF549571.1 | - | 2 - 3 | 351 | 50 | |
EF549572.1 | - | 2 - 3 | 273 | 50 | |
EU072083.1 | - | 2 - 3 | 812 | 50 | |
EU072084.1 | - | 2 - 3 | 812 | 50 | |
EU072085.1 | - | 2 - 3 | 351 | 50 | |
EU072087.1 | - | 3 - 4 | 563 | 50 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | ghrelin opposite strand/antisense RNA | GHRLOS | GHRL-AS1, GHRLAS, NCRNA00068 | Anti-sense | Chr. - 10280952 - 10293449 on Build GRCh38 | |
Downstream | SEC13 homolog, nuclear pore and COPII coat complex component | SEC13 | D3S1231E, SEC13L1, SEC13R, npp-20 | 9.9kb | Sense | Chr. - 10300929 - 10321188 on Build GRCh38 |