Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GHRL
Entrez Gene ID
51738
Gene Name
ghrelin/obestatin prepropeptide
Gene Aliases
MTLRP
Chromosome Location
Chr.3: 10285750 - 10292947 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302824.1 NP_001289753.1 1 - 2 108 66
GenBank mRNA EF549564.1 - 1 - 2 94 66
EF549572.1 - 1 - 2 78 66


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ghrelin opposite strand/antisense RNA GHRLOS GHRL-AS1, GHRLAS, NCRNA00068 Anti-sense Chr. - 10280952 - 10293449 on Build GRCh38
Downstream SEC13 homolog, nuclear pore and COPII coat complex component SEC13 D3S1231E, SEC13L1, SEC13R, npp-20 8.0kb Sense Chr. - 10300929 - 10321188 on Build GRCh38

View Full Product Details