Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BPIFA2
- Entrez Gene ID
-
140683
- Gene Name
- BPI fold containing family A member 2
- Gene Aliases
-
C20orf70, PSP, SPLUNC2, bA49G10.1
- Chromosome Location
-
Chr.20: 33161848 - 33181417 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK130543.1 |
- |
1 - 1 |
1243 |
101 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105372714 |
LOC105372714 |
|
-15.5kb
|
Sense |
Chr. - 33140337 - 33160623 on Build GRCh38 |
Downstream |
BPI fold containing family A member 4, pseudogene |
BPIFA4P |
BASE |
13.4kb
|
Sense |
Chr. - 33193605 - 33210462 on Build GRCh38 |
View Full Product Details