Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- C1orf74
- Entrez Gene ID
-
148304
- Gene Name
- chromosome 1 open reading frame 74
- Gene Aliases
-
URLC4
- Chromosome Location
-
Chr.1: 209779208 - 209784730 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_152485.3 |
NP_689698.1 |
2 - 2 |
3454 |
87 |
GenBank mRNA |
AK022798.1 |
- |
1 - 1 |
345 |
87 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BM723893.1 |
- |
1 |
205 |
|
Chr. 1 - 209780165 - 209780813 [-] on Build GRCh38 |
87 |
GenBankmRNA |
AU126981.1 |
- |
1 |
345 |
|
Chr. 1 - 209780216 - 209780953 [-] on Build GRCh38 |
87 |
GenBankmRNA |
DA969331.1 |
- |
1 |
|
|
Chr. 1 - 209780145 - 209780696 [+] on Build GRCh38 |
87 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101930114 |
LOC101930114 |
|
-21.6kb
|
Sense |
Chr. - 209661359 - 209742562 on Build GRCh38 |
View Full Product Details